Learn More
SRGAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Code produit 16811965 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16811965 20 μL 20µL
16801965 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16811965 Fournisseur Proteintech Code fournisseur 225191AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
This locus encodes a member of the SLIT-ROBO Rho GTPase activating protein family. The encoded protein stimulates GTPase activity of Rac1, and plays a role in cortical neuron development. This locus has several paralogs on human chromosome 1 resulting from segmental duplication. While this locus itself is conserved among various species, the paralogs are found only in the genus Homo, and not in the genomes of non-human great apes.
Spécification
Antigène SRGAP2
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression SRGAP2
Numéro d’ordre du gène O75044, Q91Z67
Alias de gène ARHGAP34, FNBP2, Formin binding protein 2, KIAA0456, SRGAP2, srGAP3
Symboles de gène(s) SRGAP2
Espèces hôtes Rabbit
Immunogène SRGAP2 Fusion Protein Ag18278
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 14270, 23380
Espèces cibles Mouse, Human
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG