missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ST3GAL1 Polyclonal Antibody

Code produit 15747051 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15747051 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15747051 Fournisseur Invitrogen™ Code fournisseur PA559190

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: MVLELSENLK RLIKHRPCTC THCIGQRKLS AWFDERFNQT MQPLLTAQNA LLEDDTYRWW LRLQREKKPN NLNDTIKELF RVVPGNVDPM LEKRSVGCRR CAVVGNSGNL RESSYGPEID SHDFVLRMNK APTAGFEA Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.

ST3GAL1 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. It is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of ST3GAL1 protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B.The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. Correct glycosylation of the encoded protein may be critical to its sialyltransferase activity. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4B. Two transcript variants encoding the same protein have been found for this gene. Other transcript variants may exist, but have not been fully characterized yet.
TRUSTED_SUSTAINABILITY

Spécification

Antigène ST3GAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugué Unconjugated
Formule PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Expression ST3GAL1
Numéro d’ordre du gène Q11201
Alias de gène 5330418N22Rik; AI467004; Alpha 2,3-ST 1; alpha-2,3-sialyltransferase ST3Gal I-r1; alpha-2-3-sialyltransferase; beta-galactoside alpha-2,3-sialyltransferase 1; CMP-N-acetylneuraminate: [beta-galactosidase alpha-2,3] sialytransferase; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,3-sialyltransferase 1; Gal beta1,3 GalNAc alpha2,3 sialyltransferase; Gal-beta-1,3-GalNAc-alpha-2,3-sialyltransferase; Gal-NAc6S; MGC9183; putative alpha-2,3-sialyltransferase; Sialyltransferase 4A; sialyltransferase 4A (beta-galactosidase alpha-2,3-sialytransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialyltransferase); sialyltransferase 4A (beta-galactoside alpha-2,3-sialytransferase); sialyltransferase ST3Gal-I; Siat4; SIAT4A; SIAT4-A; siat4-r1; SIATFL; ST3 beta-galactoside alpha-2,3-sialyltransferase 1; ST3Gal I; st3Gal I.1; ST3GAL1; St3gal-1; st3gal1A; st3gal1-r1; ST3GAL2; ST3GalA; ST3GalA.1; ST3GalI; ST3GAL-I sialyltransferase; ST3GalIA; ST3GalIA,1; ST3O
Symboles de gène(s) ST3GAL1
Espèces hôtes Rabbit
Immunogène Recombinant protein corresponding to Human ST3GAL1. Recombinant protein control fragment (Product #RP-89952).
Méthode de purification Antigen Affinity Chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 6482
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.