Learn More
Scienceware Kit de tête d’agitation
Simplify your stirring with this Cross Magnetic Stir Bar Kit.
Marque: Scienceware 04765-00
Code nomenclature Nacres: NB.72
Informations supplémentaires : Poids : 0.27000kg
Description
- Chemically inert.
- Low friction.
- Leakproof PTFE coating.
- Kits each offer a wide variety of stir bar sizes to accomodate different glassware in the lab.
Spécification
5 Bars | |
Kit de tête d’agitation |