Learn More
IBA Lifesciences Strep-Tactin™XT Coated Microplate
Strep-Tactin™XT coated microplate is a ready-to-use microplate for assay applications
Marque:  IBA Lifesciences 2-4101-001
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01100kg
Code produit. 15503087
Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet
Voir les produits de remplacement »
Description
Description
  • Ready-to-use Strep-Tactin™XT coated microplate (12 x 8-well strips) for ligand binding assays, immuno assays, enzyme assays, or diagnostic assays under high throughput conditions.
  • Due to the high specificity and affinity (nM-pM) of the streptavidin variant Strep-Tactin™, Strep-tag™II or Twin-Strep-tag™ fusion proteins can be selectively and stably captured even from complex mixtures.
  • Biomolecules of interest are presented to interacting partners in a uniform manner resulting in reliable and reproducible assays.
  • The solid-phase, multi-well format is compatible with standard multichannel pipettes and automated plate readers.
  • Twin-Strep-tag™ fusion protein binding capacity determined with bacterial alkaline phosphatase (97 kDa) is 4 pmol for 200 µl incubation volume.
Spécification
Spécification
1 Plate
Microplate
Promotions
For Research Use Only.