missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SURF1 Polyclonal Antibody

Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (84%), Rat (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The SURF-1 protein demonstrates a vital role in the assembly of complex IV (CIV or COX) of the mitochondrial respiratory chain. Expressed in the inner mitochondrial membrane, mutations of the SURF-1 gene generally cause cytochrome c oxidase complex IV deficiency. Shortage of complex IV leads to Leigh syndrome, a severe neurological disorder. Leigh syndrome patients are usually subject to rapidly progressive encephalopathy, characterized by necrotic lesions in subcortical brain regions. SURF-1 mutations correlate to high post-implantation embryonic lethality as well as early-onset mortality of post-natal individuals. Considerable deficit in muscle strength and motor performance is also a profound and isolated defect of SURF-1 activity in skeletal muscle and liver. Heart, brain and skeletal muscle morphological abnormalities frequently occur due to SURF-1 mutations.
Spécification
Spécification
| Antigène | SURF1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.83 mg/mL |
| Conjugué | Unconjugated |
| Formule | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Expression | SURF1 |
| Numéro d’ordre du gène | Q15526 |
| Alias de gène | 0610010F23Rik; Ab1-205; CMT4K; Surf1; Surf-1; surfeit 1; surfeit gene 1; surfeit locus protein 1 |
| Symboles de gène(s) | SURF1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?