Learn More
MP Biomedicals™ T4 gp32 Bulk
Binds to single-stranded DNA in a co-operative manner, thereby destabilizing double-stranded DNA. MP Biomedicals T4 gp32 Bulk is a nonsequence specific, single strand binding protein, isolated from the bacteriophage T4.
Marque: MP Biomedicals™ 11TGP32100
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Description
The supplied protein is a purified recombinant form free of endonucleases and nickases. The components of the storage buffer are: Tris-HCl 20mm, pH 8.0, NaCl 100mm, EDTA 1mm, DTT 0.5mm, Glycerol 50%.- Enhances DNA polymerase proofreading activity and PCR fidelity
- Increases yield of large PCR DNA fragments
Spécification
100 μg | |
RUO | |
T4 bacteriophage gene 32 protein |