missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF7 Polyclonal Antibody
GREENER_CHOICE

Code produit 17287543 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17287543 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17287543 Fournisseur Invitrogen™ Code fournisseur PA5114783

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF7.

The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. The general transcription factor, TFIID, consists of the TATA-binding protein (TBP) associated with a series of TBP-associated factors (TAFs) that together participate in the assembly of the transcription preinitiation complex.
TRUSTED_SUSTAINABILITY

Spécification

Antigène TAF7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Expression TAF7
Numéro d’ordre du gène Q15545, Q9R1C0
Alias de gène 55kDa; AI607308; BB007175; RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF2F; Taf7; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII55; TAFII-55; TATA box binding protein (TBP) associated factor, RNA polymerase II, F, 55 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TATA-box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit
Symboles de gène(s) TAF7
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from human TAF7(Accession Q15545), corresponding to amino acid residues R180-S230.
Méthode de purification Affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 24074, 307485, 6879
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.