Learn More
Fisherbrand™ Tank Vent Filter
Contains activated carbon. Fisher Scientific™ Tank Vent Filter also provides membrane filtration and chemical absorption.
Marque: Fisherbrand™
Code nomenclature Nacres: JA.13
Informations supplémentaires : Poids : 0.16200kg
Description
- Prevents airborne particles, organic and bacteria from entering the PE water tank.
- Integral scavenger removes CO2
Spécification
Accu500 | |
Filter, Vent | |
CO2 scavenger |