Learn More
TEX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16827254 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16827254 20 μL 20µL
16817254 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16827254 Fournisseur Proteintech Code fournisseur 173721AP20UL
Rabbit Polyclonal Antibody
Testis-expressed sequence 10 protein (Tex10) functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex. This complex is recruited to ZNF148 by methylated CHTOP leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes. Tex10 is also involved in chromatin binding, maturation of LSU-rRNA, regulation of DNA-dependent DNA replication initiation, and rRNA processing.
Spécification
Antigène TEX10
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression TEX10
Numéro d’ordre du gène Q3URQ0, Q9NXF1
Alias de gène Nbla10363, testis expressed 10, TEX10
Symboles de gène(s) TEX10
Espèces hôtes Rabbit
Immunogène TEX10 Fusion Protein Ag11014
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 269536, 298065, 54881
Espèces cibles Mouse, Human, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG