Learn More
Invitrogen™ Tigatuzumab Recombinant Monoclonal Antibody
GREENER_CHOICE
Code produit 30284547
Change view
Click to view available options
Quantité:
1 mg
100 μg
Conditionnement:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30284547 100 μg 100µg
30284522 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30284547 Fournisseur Invitrogen™ Code fournisseur MA558303
Recombinant Monoclonal Antibody
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Tigatuzumab is under investigation in clinical trial NCT00991796, CS-1008 With Carboplatin/Paclitaxel in Chemotherapy naive Subjects With Metastatic or Unresectable Non-small Cell Lung Cancer (NSCLC).
TRUSTED_SUSTAINABILITY
Spécification
Antigène Tigatuzumab Humanized
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Alias de gène CS-1008
Espèces hôtes Human
Méthode de purification Protein A
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Lyophilized
Isotype IgG1