Learn More
Lovibond™ Tintometer™ Combipack Molybdate LR, Molybdate LR No. 1 / No. 2
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marque: Lovibond™ 517772
Code nomenclature Nacres: NB.61
Informations supplémentaires : Poids : 0.30000kg
Description
- Combi-Pack Molybdate LR
- Molybdate LR No. 1and Molybdate LR No. 2
- Available as two packs of 100 tablets or two packs of 250
Spécification
Photometry Tablet Reagents | |
2 Packs of 250 Tablets | |
Combipack Molybdate LR, Molybdate LR No. 1 / No. 2 |
Test kits, colorimeters and spectrophotometers | |
250 x Molybdate LR No. 1; 250 x Molybdate LR No. 2 Tablets |