Learn More
Abnova™ TNFRSF7 (Human) Recombinant Protein
Human TNFRSF7 full-length ORF ( AAH12160, 1 a.a. - 260 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000939-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MARPHPWWLCVLGTLVGLSATPAPKSCPERHYWAQGKLCCQMCEPGTFLVKDCDQHRKAAQCDPCIPGVSFSPDHHTRPHCESCRHCNSGLLVRNCTITANAECACRNGWQCRDKECTECDPLPNPSLTARSSQALSPHPQPTHLPYVSEMLEARTAGHMQTLADFRQLPARTLSTHWPPQRSLCSSDFIRILVIFSGMFLVFTLAGALFLHQRRKYRSNKGESPVEPAEPCRYSCPREEEGSTIPIQEDYRKPEPACSP
Spécification
AAH12160 | |
CD27 molecule | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD27 | |
GST |
939 | |
Wheat germ expression system | |
25 μg | |
MGC20393, S152, T14, TNFRSF7, Tp55 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |