missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TPX2 Monoclonal Antibody (18D5-1)
GREENER_CHOICE

Code produit 11525422
Change view
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11525422 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11525422 Fournisseur Invitrogen™ Code fournisseur MA1802

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

MA1-802 detects the TPX2 in human, rat and mouse cells. MA1-802 has been successfully used in Western blot, immunoprecipitation and immunofluorescence procedures. By Western blot, this antibody detects a 92 kDa protein. The MA1-802 immunogen is recombinant human TPX2.

The protein encoded by this gene is found in the nucleolus and is similar to that of a family of genes on the Y-chromosome. This gene is intronless. Defects in this gene are a cause of sudden infant death with dysgenesis of the testes syndrome (SIDDT).
TRUSTED_SUSTAINABILITY

Specifications

Antigène TPX2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 18D5-1
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1mg/mL BSA and 0.05% sodium azide
Expression Tpx2
Numéro d’ordre du gène A2APB8, Q9ULW0
Alias de gène 2610005B21Rik; C20orf1; C20orf2; Differentially expressed in cancerous and non-cancerous lung cells 2; differentially expressed in lung cells; DIL2; DIL-2; FLS353; GD: HCA519; GD:C20orf1; HCA519; HCTP4; Hepatocellular carcinoma-associated antigen 519; hepatocellular carcinoma-associated antigen 90; p100; preferentially expressed in colorectal cancer; protein fls353; REPP86; restricted expression proliferation associated protein 100; restricted expression proliferation-associated protein 100; Targeting protein for Xklp2; TPX 2; Tpx2; TPX2 microtubule nucleation factor; TPX2, microtubule nucleation factor; TPX2, microtubule-associated; TPX2, microtubule-associated protein homolog; TPX2, microtubule-associated protein homolog (Xenopus laevis); TPX2, microtubule-associated, homolog
Symboles de gène(s) Tpx2
Espèces hôtes Mouse
Immunogène Recombinant human TPX2.
Méthode de purification Protein G
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 22974, 311546, 72119
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.