Learn More
MedChemExpress Trastuzumab
Code produit 16402288
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16402288 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16402288 Fournisseur MedChemExpress Code fournisseur HYP9907
MedChemExpress Trastuzumab is a humanized IgG1 monoclonal antibody for patients with invasive breast cancers that overexpress HER2. Trastuzumab has the potential for HER2 Positive Metastatic Breast Cancer and HER2 Positive Gastric Cancer research.
Spécification
Antigène EGFR
Quantité 5 mg
Espèces cibles Human
Contenu et stockage Please store the product under the recommended conditions in the Certificate of Analysis.
Type de produit Inhibitory Antibodies
Conditions d’expédition Room temperature
CAS 180288-69-1
Pureté ou qualité 99.2%