missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC22D1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Tous les produits Proteintech ProduitsDescription
This antibody is a rabbit polyclonal antibody raised against an internal region of human TSC22D1.
TSC22 domain family protein, or TSC22, is a transcription factor that belongs to the large family of early response genes. This transcriptional repressor is known to act on the C-type natriuretic peptide (CNP) promoter. TSC22 belongs to the TSC-22/Dip/Bun family and is an intracellular protein that may be found in the cytoplasm or nucleus. This transcription factor is known to regulate cell growth, differentiation and cell death and is involved in modulating the transcriptional activity of Smad3 and Smad4. This protein is ubiquitously expressed in most tissues and widely in both fetal and adult tissues. It is generally expressed in aortic endothelial cells, and induced by cytokines, including TGFB. These proteins may be possible therapeutic targets of leukemia and prostate cancer.
Spécification
Spécification
| Antigène | TSC22D1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Expression | TSC22D1 |
| Numéro d’ordre du gène | P62500, P62501, Q15714 |
| Alias de gène | Cerebral protein 2, KIAA1994, Regulatory protein TSC 22, RP11 269C23.2, TGFB1I4, TSC22, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22D1 |
| Symboles de gène(s) | Tsc22d1 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?