Learn More
Invitrogen™ TWEAK Monoclonal Antibody (189803)
GREENER_CHOICE
Code produit 30534561
Change view
Click to view available options
Quantité:
500 μg
Conditionnement:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30534561 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30534561 Fournisseur Invitrogen™ Code fournisseur MA560950
Rat Monoclonal Antibody
Reconstitute at 0.5 mg/mL in sterile PBS.
TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.
TRUSTED_SUSTAINABILITY
Spécification
Antigène TWEAK
Applications ELISA
Classification Monoclonal
Clone 189803
Concentration 0.5 mg/mL
Conjugué Unconjugated
Formule PBS with 5% trehalose and no preservative
Expression Tnfsf12
Numéro d’ordre du gène O54907
Alias de gène APO3 ligand; APO3/DR3 ligand; APO3L; Dr3l; Dr3lg; LOW QUALITY PROTEIN: tumor necrosis factor ligand superfamily member 12; MGC129581; MGC20669; TNF superfamily member 12; TNF-related weak inducer of apoptosis; TNFSF12; TNLG4A; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4A; tumor necrosis factor ligand superfamily member 12; Tumor necrosis factor ligand superfamily member 12, membrane form; Tumor necrosis factor ligand superfamily member 12, secreted form; tumor necrosis factor superfamily member 12; TWEAK; UNQ181/PRO207; zgc:153941
Symboles de gène(s) Tnfsf12
Espèces hôtes Rat
Immunogène E. coli-derived recombinant mouse TWEAK/TNFSF12, Arg105-His249.
Méthode de purification Protein A/G
Quantité 500 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 21944
Espèces cibles Mouse
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Lyophilized
Isotype IgG2a