Learn More
UGT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Code produit 16878004 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16878004 20 μL 20µL
16868004 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16878004 Fournisseur Proteintech Code fournisseur 179821AP20UL
Rabbit Polyclonal Antibody
This antibody specifically recognizes the endogenous UGT8.
UGT8 (UDP-galactose-ceramide galactosyltransferase, 2-hydroxyacylsphingosine 1-beta-galactosyltransferase) is a 541 amino acid, single pass membrane protein of the UDP-glycosyltransferase family. UGT8 is believed to be primarily involved with the metabolism of sphingolipids and galactosylceramide biosynthesis. UGT8 is one of six genes whose elevated expression has been correlated with a significantly increased the risk of lung metastases in breast cancer patients. As such, UGT8 may be a significant index of tumor aggressiveness and a potential marker for the prognostic evaluation of lung metastases in breast cancer. UGT8 is ubiquitously expressed with highest levels found in central and peripheral nervous systems and is up-regulated in breast cancers.
Spécification
Antigène UGT8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Expression UGT8
Numéro d’ordre du gène Q09426, Q16880, Q64676
Alias de gène ceramide galactosyltransferase, Cerebroside synthase, CGT, UDP glycosyltransferase 8, UGT4, UGT8
Symboles de gène(s) Ugt8
Espèces hôtes Rabbit
Immunogène UGT8 Fusion Protein Ag12393
Méthode de purification Antigen Affinity Chromatography
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 22239, 50555, 7368
Espèces cibles Rat, Human, Mouse
Contenu et stockage -20°C
Type de produit Antibody
Forme Liquid
Isotype IgG