missing translation for 'onlineSavingsMsg'
Learn More

Vγ 3 TCR Hamster anti-Mouse, BUV395, Clone: 536, BD Optibuild™

Code produit 15838119
Change view
Click to view available options
Quantité:
50 μg
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15838119 50 μg 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15838119 Fournisseur BD Biosciences Code fournisseur 743244

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Edge
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Syrian Hamster Monoclonal Antibody

The 536 monoclonal antibody specifically recognizes Vγ3† T-cell Receptor (TCR)-bearing T lymphocytes, which are the predominantγδTCR-bearing cells in the early fetal thymus and the adult epidermis of euthymic mice. The first T cells to mature in the embryonic thymus express the Vγ3 and Vδ1 TCR chains, and their development is dependent upon IL-7. There is evidence that the Vγ3 TCR-bearing fetal thymocytes are the precursors of the majority of dendritic epidermal T cells (DEC), which may be replenished in the adult by proliferation in situ /rather than by seeding from primary lymphoid organs. Although the Vγ3 TCR is almost exclusively found in the DEC population, it has been shown that the homing of DEC to the epidermis does not require expression of the Vγ3 gene segment. Vγ3 TCR-bearing dermal dendritic cells have also been described. Vγ3 TCR has also been found on a subset of T lymphocytes in the lactating mammary gland and at the site of antigenic challenge in contact-sensitized mice. Plate-bound 536 antibody activates Vγ3 TCR-bearing T cells, and Fab fragments of mAb 536 block in vitro /stimulation of DEC by keratinocytes.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348 nm and an Em Max at 395 nm, BD Horizon BUV395 can be excited with a 355 nm laser and detected with a 379/28 filter. BUV395 is a UV-excitable dye that has been developed exclusively by BD Biosciences. With an excitation max of 348 nm and emission max of 395 nm, BUV395 can be excited by the 355nm laser and detected in a 379/28 filter. This dye is optimal for multicolor flow cytometry because it has little to no spillover into other detectors.
TRUSTED_SUSTAINABILITY

Spécification

Antigène Vγ 3 TCR
Classification Monoclonal
Clone 536
Concentration 0.2mg/mL
Conjugué Brilliant Ultraviolet 395
Formule Aqueous buffered solution containing ≤0.09% sodium azide.
Alias de gène TCR Vγ3, TCR V gamma 3, TCR Vg3
Espèces hôtes Syrian Hamster
Immunogène AKR mouse dendritic epidermal cell clone 7-17
Méthode de purification Affinity Purified
Quantité 50 μg
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Mouse
Contenu et stockage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.