Learn More
Novus Biologicals™ Recombinant Human CaM Kinase II alpha GST (N-Term) Protein
Tous les produits Bio Techne Produits
Click to view available options
Quantité:
10 μg
25 μg
Conditionnement:
10µg
25µg
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Spécification
Spécification
| À utiliser avec (application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
| Formule | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
| Identification génétique (Entrez) | 815 |
| Poids moléculaire | M.W. (Theoretical): 37.29 kDa |
| Quantité | 25 μg |
| Conditions de stockage | Store at -80C. Avoid freeze-thaw cycles. |
| Alias de gène | calcium/calmodulin-dependent protein kinase (CaM kinase) II alpha, calcium/calmodulin-dependent protein kinase II alpha, calcium/calmodulin-dependent protein kinase II alpha-B subunit, CaM kinase II alpha subunit, CaM kinase II subunit alpha, CAMKAcalcium/calmodulin-dependent protein kinase type II subunit alpha, CaMK-II alpha subunit, CaMK-II subunit alpha, CaMKIINalpha, CaM-kinase II alpha chain, EC 2.7.11, EC 2.7.11.17, KIAA0968calcium/calmodulin-dependent protein kinase type II alpha chain |
| Symbole de gène(s) | CAMK2A |
| Catégorie de recherche | Protein Kinase, Wnt Signaling Pathway |
| Pureté ou qualité | >80% by SDS-PAGE and Coomassie blue staining |
| Afficher plus |
Correction du contenu d'un produit
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido