Learn More
Goat anti-Ferret IgG (H+L) Secondary Antibody, Allophycocyanin, Novus Biologicals™
Goat Polyclonal Antibody
Marque:  Novus Biologicals NB7222APC
Code nomenclature Nacres: NA.46
Code produit. 30230814
  • 508.00€ / 1mL
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Ferret samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
IgG (H+L)
Polyclonal
PBS
This Goat anti-Ferret IgG (H+L) Secondary Antibody was developed against ferret IgG-heavy and light chain.
1 mL
Adaptive Immunity, Immunology
Ferret
Purified
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin)
APC
Goat
Immunogen affinity purified
RUO
Secondary
Store at 4°C in the dark.
IgG