missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WT1 Polyclonal Antibody
GREENER_CHOICE

Code produit 16342045
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
16342045 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 16342045 Fournisseur Invitrogen™ Code fournisseur PA585096

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (97%), Japanese Medaka (83%), Xenopus laevis (87%), Pig (97%), Chicken (94%). Positive Control: Mouse testis, Rat testis. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The Wilms Tumor 1 gene encodes one of the WPP domain-interacting proteins (Wilms Tumor 1/AT5G11390, WIT2/AT1G68910) required for RanGAP nuclear envelope association in root tip cells. Ran GTPase plays essential roles in multiple cellular processes, including nucleocytoplasmic transport, spindle formation, and postmitotic nuclear envelope reassembly. The cytoplasmic Ran GTPase activating protein RanGAP is critical to establish a functional RanGTP/RanGDP gradient across the nuclear envelope and is associated with the outer surface of the nuclear envelope in metazoan and higher plant cells. Arabidopsis thaliana RanGAP association with the root tip nuclear envelope requires a family of likely plant-specific nucleoporins combining coiled-coil and transmembrane domains (CC-TMD) and WPP domain-interacting proteins (WIPs). Wilms Tumor 1 and WIT2 have been identified as a second family of CC-TMD proteins, structurally similar, yet clearly distinct from the WIP family, that is required for RanGAP nuclear envelop association in root tip cells.
TRUSTED_SUSTAINABILITY

Spécification

Antigène WT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugué Unconjugated
Formule PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Expression WT1
Numéro d’ordre du gène P19544, P22561, P49952
Alias de gène AWT1; D630046I19Rik; EWS-WT1; GUD; NPHS4; WAGR; Wilms tumor 1; Wilms tumor 1 homolog; Wilms tumor protein; Wilms tumor protein homolog; Wilms tumor protein isoform Ex4a(+); WIT1; WIT-1; WIT-2; WT; Wt1; Wt-1; WT1 antisense RNA; WT1AS; WT1-AS; WT1-AS1; WT33
Symboles de gène(s) WT1
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the center region of human Wilms Tumor 1. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 22431, 24883, 7490
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.