Learn More
Waters Corporation XBridge Prep Shield RP18 Column
XBridge BEH Shield RP18 columns complement the XBridge BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development.
Marque:  Waters Corporation 186003991
Informations supplémentaires : Poids : 0.50000kg
Code produit. 13273300
  • 772.50€ / 1 pièce
Expédition estimée: 11-07-2024
Description
The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Spécification
Spécification
19 mm
6000 psi (415 Bar)
10 μm
1/Ea.
Yes
10 mm
Spherical
130 Å
L1
Column