Learn More
YAP1 Mouse anti-Human, Unconjugated, Clone: 3A7A9, Proteintech
Code produit 16899175 Tous les produits Proteintech Produits
Modifier l'affichage
Click to view available options
Quantité:
20 μL
150 μL
Conditionnement:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
16899175 150 μL 150µL
16809185 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16899175 Fournisseur Proteintech Code fournisseur 669001IG150UL
 Article restreint
En stock
Ajouter au panier
Edge
Ajouter au panier
Mouse Monoclonal Antibody
The YAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YAP1. This antibody recognizes human, mouse, rat antigen. The YAP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Antigène YAP1
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3A7A9
Concentration 1000 μg/mL
Conjugué Unconjugated
Espèces hôtes Mouse
Quantité 150 μL
Primaire ou secondaire Primary
Identification génétique (Entrez) 10413
Espèces cibles Human, Rat, Mouse
Isotype IgG1