Learn More
Complement C3 Antibody (F1-8), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque:  Novus Biologicals NBP2-46609MFV450
Code nomenclature Nacres: NA.46
599.25 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Code produit. 18705120
  • 748.00€ / 0.10mL
Description
Complement C3 Monoclonal antibody specifically detects Complement C3 in Human samples. It is validated for ELISA, Immunoprecipitation, Affinity Purification
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Complement C3
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
ELISA, Immunoprecipitation, Affinity Purification
F1-8
ELISA, Immunoprecipitation, Immunoaffinity Purification
Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3
This Complement C3 Antibody (F1-8) was developed against Complement component 3 (C3) purified from human plasma.
0.1 mL
Angiogenesis, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Mesenchymal Stem Cell Markers, Neurodegeneration, Neuroscience, Signal Transduction
718
Store at 4C in the dark.
IgG1 κ