Learn More
USAG1/SOSTDC1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-99425JF646
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
USAG1/SOSTDC1 Polyclonal antibody specifically detects USAG1/SOSTDC1 in Mouse samples. It is validated for Western Blot
Spécification
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Mouse USAG1/SOSTDC1. | |
| 25928 | |
| Purified |
| Primary | |
| Store at 4C in the dark. | |
| IgG |