Learn More
Hamilton™ 10 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Marque:  Hamilton™ CAL80508
Informations supplémentaires : Poids : 0.04500kg
Code produit. 17864332
  • 328.00€ / 1 pièce
Expédition estimée: 02-07-2024
Description
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 10 μL.
Spécification
Spécification
Not Autoclavable
26s G
0.005 in.
0.0187 in.
0.485 mm
0.47 mm
122°F
50°F
10°C
10 μL
Cone
Borosilicate Glass
0.019 in.
0.13 mm
0.260 in.
6.60 mm
2 in.
50°C
701 N
1
51 mm
Cemented