Learn More
FGFR2 Antibody (2E9), Novus Biologicals™
Code produit 30043495 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μL
50 μL
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30043495 100 μL 100µL
30043955 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30043495 Fournisseur Novus Biologicals Code fournisseur NBP326583100ul
Rabbit Monoclonal Antibody
FGFR2 Monoclonal antibody specifically detects FGFR2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Spécification
Antigène FGFR2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2E9
Conjugué Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formule PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gène BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from Human FGFR2 [UniProt P21802]
Méthode de purification Affinity purified
Quantité 100 μL
État réglementaire RUO
Disciplines de recherche Angiogenesis, Protein Kinase
Primaire ou secondaire Primary
Identification génétique (Entrez) 2263
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at -20 to -70C. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG