missing translation for 'onlineSavingsMsg'
Learn More

Insulin R/CD220 Antibody (INSR/2277R), Alexa Fluor™ 488, Novus Biologicals™

Code produit 18529454 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18529454 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18529454 Supplier Novus Biologicals Supplier No. NBP308838AF488

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Insulin R/CD220 Monoclonal antibody specifically detects Insulin R/CD220 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigène Insulin R/CD220
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone INSR/2277R
Conjugué Alexa Fluor 488
Formule 50mM Sodium Borate
Alias de gène CD 220, CD220, CD220 antigen, EC 2.7.10, EC 2.7.10.1, HHF5, insulin receptor, IR
Espèces hôtes Rabbit
Immunogène Recombinant fragment of extracellular domain of human Insulin Receptor alpha (exact sequence is proprietary) (Uniprot: P06213)
Méthode de purification Protein A or G purified
Quantité 100 μg
État réglementaire RUO
Disciplines de recherche Diabetes Research, Lipid and Metabolism, Neuroscience, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primaire ou secondaire Primary
Identification génétique (Entrez) 3643
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.