missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 14F7 Recombinant Mouse Monoclonal Antibody

Mouse Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA541734
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
14F7 is a biosimilar that targets ganglioside N-glycolyl GM3. GM2 and GD2 gangliosides are sialic acid-containing glycosphingolipids. GalNAc-T is the enzyme involved in the biosynthesis of G(M2) and G(D2) glycosphingolipids. GalNAc-T catalyzes the transfer of GalNAc into G(M3) and G(D3) by a beta-1,4 linkage, resulting in the synthesis of G(M2) and G(D2), respectively. Three transcript variants encoding different isoforms have been found for this gene.
Spécification
| 14F7 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| 14F7 | |
| Human ganglioside N-glycolyl GM3. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, SDS-Page | |
| 2.9 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu