Learn More
Hamilton™ 25 μL 700 Series Microliter Syringe
The 700 Series Microliter Syringe fulfills the general liquid handling requirements of manufacturing and research laboratories.
Marque:  Hamilton™ CAL84855
Informations supplémentaires : Poids : 0.07100kg
Code produit. 17804202
  • 349.00€ / 1 pièce
Expédition estimée: 02-07-2024
Description
  • The 700 series is the original Hamilton syringe.
  • This series of Microliter syringes is designed to solve the general liquid handling requirements of manufacturing and research laboratories.
  • The 700 series remains the industry standard.
  • The plungers and barrels are not interchangeable or replaceable.
  • Dispenses volumes up to 25 μL.