missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 3-Nitrotyrosine Monoclonal Antibody (2A12)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA112770
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
3-Nitrotyrosine Monoclonal Antibody for Western Blot
Protein tyrosine nitration results in a post-translational modification that is increasingly receiving attention as an important component of nitric oxidesignaling. While multiple nonenzymatic mechanisms are known to be capable of producing nitrated tyrosine residues, most tyrosine nitration events involve catalysis by metalloproteins such as myeloperoxidase, eosinophilperoxidase, myoglobin, the cytochrome P-450s, superoxide dismutase and prostacyclin synthase. Various studies have shown that protein tyrosinenitration is limited to specific proteins and that the process is selective. For example, exposure of human surfactant protein A (SP-A) to oxygen-nitrogen intermediates generated by activated alveolar macrophages resulted in specific nitration of SP-A at tyrosines 164 and 166, while addition of 1.2 mMCO 2 resulted in additional nitration at tyrosine 161. The presence of nitrotyrosine-containing proteins has shown high correlation to disease states such as atherosclerosis, Alzheimer's disease, Parkinson's disease and amyotrophic lateral sclerosis.
Spécification
| 3-Nitrotyrosine | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 3mM sodium azide; pH 7.5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 2A12 | |
| Unconjugated | |
| 3-Nitro-tyrosine; C9H10N2O5 | |
| 3-Nitrotyrosine-KLH. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu