missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SMCX Polyclonal antibody specifically detects SMCX in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigène | SMCX |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | DyLight 680 |
| Formule | 50mM Sodium Borate |
| Alias de gène | DXS1272EMRXJ, EC 1.14.11, EC 1.14.11.-, Histone demethylase JARID1C, JARID1Clysine-specific demethylase 5C, jumonji, AT rich interactive domain 1C, Jumonji, AT rich interactive domain 1C (RBP2-like), Jumonji/ARID domain-containing protein 1C, lysine (K)-specific demethylase 5C, MRXSJ, Protein SmcX, Protein Xe169, Smcx homolog, X chromosome, SMCXselected cDNA on X, Smcy homolog, X-linked, Smcy homolog, X-linked (mouse), XE169JmjC domain-containing protein SMCX |
| Espèces hôtes | Rabbit |
| Immunogène | A synthetic peptide corresponding to a sequence within amino acids 1400 to the C-terminus of human SMCX (XP_011529128.1).,, Sequence:, ERHGSRARGRALERRRRRKVDRGGEGDDPAREELEPKRVRSSGPEAEEVQEEEELEEETGGEGPPAPIPTTGSPSTQENQNGLEPAEGTTSGPSAPFSTLTPRLHLPCPQQPPQQQL |
| Méthode de purification | Affinity purified |
| Quantité | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?