missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody (LOB12), Alexa Fluor™ 532, Novus Biologicals™
Rat Monoclonal Antibody
Marque: Novus Biologicals NB100-63415AF532
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Mouse samples. It is validated for Flow Cytometry
Spécification
| 4-1BB/TNFRSF9/CD137 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| Murine CD137 human Fc fusion protein | |
| 0.125 mL | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
| 3604 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| LOB12 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu