missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A2BP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-98015-100ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
A2BP1 Polyclonal antibody specifically detects A2BP1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Spécification
| A2BP1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:500-1:5000 | |
| 2BP1, A2BP, A2BP1 Hexaribonucleotide-binding protein 1, Ataxin-2-binding protein 1, FOX-1, FOX1 hexaribonucleotide binding protein 1, Fox-1 homolog A, HRNBP1 ataxin 2-binding protein 1, RNA binding protein fox-1 homolog 1, RNA binding protein, fox-1 homolog (C. elegans) 1 | |
| Produced in rabbits immunized with E. coli-derived Human A2BP1 fragment. | |
| 100 μg | |
| Epigenetics, Neuroscience | |
| 54715 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu