missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABCB4 Monoclonal Antibody (P3II-26), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA135605
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The membrane-associated protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MDR/TAP subfamily. Members of the MDR/TAP subfamily are involved in multidrug resistance. The protein encoded by this gene is an ATP-dependent drug efflux pump for xenobiotic compounds with broad substrate specificity. It is responsible for decreased drug accumulation in multidrug-resistant cells and often mediates the development of resistance to anticancer drugs. This protein also functions as a transporter in the blood-brain barrier.
Spécification
| ABCB4 | |
| Monoclonal | |
| Unconjugated | |
| P21439 | |
| ABCB4 | |
| MDR3. | |
| RUO | |
| 5244 | |
| 2-8°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
| P3II-26 | |
| ABCB4 | |
| ABC20, CD243, CLCS, GP170, MDR1, MGC163296, P-GP, PGY1 | |
| Mouse | |
| 1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu