missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACACB Polyclonal Antibody

Goat Polyclonal Antibody
Marque: Invitrogen™ PA5142810
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
Spécification
| ACACB | |
| Polyclonal | |
| Unconjugated | |
| ACACB | |
| Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 32 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O00763 | |
| ACACB | |
| Peptide with sequence C-TLNVQESGRVKYIKR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu