Learn More
Invitrogen™ ACE Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA596210
Description
Immunogen sequence: LHQCDIYRST KAGAKLRKVL QAGSSRPWQE VLKDMVGLDA LDAQPLLKYF QPVTQWLQEQ NQQNGEVLGW PEYQWHPPLP DNYPEGIDLV TDEAEASKFV EEYDRTSQVV WNEYAEANWN YNTNITTETS KILLQKNMQI ANHTLKYGTQ ARKFDVNQLQ NTTIKRIIKK VQDLERAALP AQELEEYNKI LLDMETTYSV ATVCH Positive Samples: HepG2, MCF7, Mouse lung; Cellular Location: Cell membrane, Cytoplasm, Single-pass type I membrane protein.
This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287 bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes-the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined.
Spécification
| ACE | |
| Polyclonal | |
| Unconjugated | |
| Ace | |
| ACE; ACE1; ACE-T; angiotensin 1 converting enzyme 1; angiotensin converting enzyme; angiotensin converting enzyme, somatic isoform; angiotensin I converting enzyme; angiotensin I converting enzyme (peptidyl-dipeptidase A) 1; angiotensin I converting enzyme 1; angiotensin I converting enzyme peptidyl-dipeptidase A 1 transcript; angiotensin I-converting enzyme (Dipeptidyl carboxypeptidase 1); angiotensin-converting enzyme; Angiotensin-converting enzyme, soluble form; angiotensin-converting enzyme; LOW QUALITY PROTEIN: angiotensin-converting enzyme; angiotensin-converting enzyme-like; AW208573; carboxycathepsin; CD143; CD143 antigen; DCP; Dcp1; dipeptidyl carboxypeptidase 1; Dipeptidyl carboxypeptidase 1 (Angiotensin I-converting enzyme); dipeptidyl carboxypeptidase I; dipeptidyl peptidase; I79_009670; ICH; kininase II; LOC103158533; MGC26566; MVCD3; peptidase P; peptidyl dipeptidase i; StsRR92; testicular ECA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11421, 1636, 24310 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P09470, P12821, P47820 | |
| Ace | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 554-758 of human ACE11 (NP_0007801). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.