missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetyl-Tubulin alpha-1B (Lys40) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA5101039
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of Acetyl-alpha-Tubulin only when acetylated at Lys40.
The microtubules are intracellular dynamic polymers made up of evolutionarily conserved polymorphic alpha/beta-tubulin heterodimers and a large number of microtubule-associated proteins (MAPs). Microtubules have their intrinsic polarity; highly dynamic plus ends and less dynamic minus ends. Microtubules are required for vital processes in eukaryotic cells including mitosis, meiosis, maintenance of cell shape and intracellular transport. Microtubules are also necessary for movement of cells by means of flagella and cilia. In mammalian tissue culture cells microtubules have their minus ends anchored in microtubule organizing centers (MTOCs).The GTP (guanosintriphosphate) molecule is an essential for tubulin heterodimer to associate with other heterodimers to form microtubule. In vivo, microtubule dynamics vary considerably. Microtubule polymerization is reversible and a populations of microtubules in cells are on their minus ends either growing or shortening - this phenomenon is called dynamic instability of microtubules. The alpha-tubulin (relative molecular weight around 50 kDa) is globular protein that exists in cells as part of soluble alpha/beta-tubulin dimer or it is polymerized into microtubules. In different species it is coded by multiple tubulin genes that form tubulin classes (in human 6 genes).
Spécification
| Acetyl-Tubulin alpha-1B (Lys40) | |
| Polyclonal | |
| Unconjugated | |
| TUBA1B | |
| alpha tubulin; alpha-tubulin 2; Alpha-tubulin isotype M-alpha-2; alpha-tubulin ubiquitous; Detyrosinated tubulin alpha-1B chain; K-ALPHA-1; RGD1565476; Tuba1b; Tuba2; tubulin alpha 1b; tubulin alpha-1B chain; Tubulin alpha-2 chain; tubulin alpha-ubiquitous chain; Tubulin K-alpha-1; tubulin, alpha 1B; tubulin, alpha 2; tubulin, alpha, ubiquitous | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10376, 22143, 500929 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05213, P68363, Q6P9V9 | |
| TUBA1B | |
| A synthesized peptide derived from human TUBA1B(Accession P68363), corresponding to amino acid residues L23-F52. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu