Learn More
Activin RIIB Antibody (bimagrumab), CoraFluor™ 1, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals NBP3-28282CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Activin RIIB Monoclonal antibody specifically detects Activin RIIB in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| Activin RIIB | |
| Monoclonal | |
| CoraFluor 1 | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActR-IIB, ACTRIIB, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| ACVR2B | |
| 0.1 mL | |
| Protein Kinase | |
| 93 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| bimagrumab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |