missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AF4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP1-28705CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AF4 Polyclonal antibody specifically detects AF4 in Human samples. It is validated for Immunoprecipitation,Western Blot (Negative)
Spécification
| AF4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot (Negative) | |
| CoraFluor 1 | |
| AF-4, AF4/FMR2 family member 1, AF4/FMR2 family, member 1, AF4myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 2, ALL1-fused gene from chromosome 4 protein, FEL, MLLT2MGC134969, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 2, PBM1, pre-B-cell monocytic leukemia partner 1, Protein AF-4, Protein FEL, Proto-oncogene AF4 | |
| The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human proto-oncogene AF4 using the numbering given in entry NP_005926.1 (GeneID 4299). | |
| 0.1 mL | |
| Cancer | |
| 4299 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu