missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AFP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA516658
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PA5-16658 targets Alpha Fetoprotein in IHC (P) and WB applications and shows reactivity with Human, Ovine, and Porcine samples. The PA5-16658 immunogen is purified alpha-fetoprotein.
AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms.
Spécification
| AFP | |
| Polyclonal | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P02771, Q8MJ76 | |
| AFP | |
| Purified alpha-fetoprotein | |
| 500 μL | |
| Primary | |
| Human, Ovine, Pig | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| AFP | |
| a fetoglobulin; a fetoprotein; Afp; AFPD; Alpha fetoglobulin; alpha fetoprotein; alpha-1-fetoprotein; Alpha-fetoglobulin; alpha-fetoprotein; alpha-fetoprotein precursor; alpha-foetoprotein; chloride intracellular channel 6; FETA; fetoglobulin; fetoprotein; HPAFP; a fetoglobulin; a fetoprotein | |
| Rabbit | |
| purified | |
| RUO | |
| 101103646, 174, 397586 | |
| 4° C | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu