missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AGER Antibody (XT-M4) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-28638-1mg
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
AGER Monoclonal antibody specifically detects AGER in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Spécification
| AGER | |
| Monoclonal | |
| Unconjugated | |
| Q15109 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| XT-M4 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| advanced glycosylation end product-specific receptor, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products | |
| AGER/ RAGE | |
| 1 mg | |
| Cancer | |
| 177 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu