Learn More
AlaRS Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67909-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The AlaRS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human AlaRS. This antibody recognizes human, mouse, rat antigen. The AlaRS antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Spécification
| AlaRS | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 16 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 1C8H3 | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 107 kDa | |
| Liquid |