missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlaRS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17394-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The human alanyl-tRNA synthetase (AARS) belongs to a family of tRNA synthases,of the class II enzymes.Class II tRNA synthases evolved early in evolution and are highly conserved.This is reflected by the fact that 498 of the 968-residue polypeptide human AARS shares 41% identity witht the E.coli protein.tRNA synthases are the enzymes that interpret the RNA code and attach specific aminoacids to the tRNAs that contain the cognate trinucleotide anticodons.They consist of a catalytic domain which interacts with the amino acid acceptor-T psi C helix of the tRNA, and a second domain which interacts with the rest of the tRNA structure.Spécification
| AlaRS | |
| Polyclonal | |
| Unconjugated | |
| AARS | |
| AARS, Alanine tRNA ligase, alanyl tRNA synthetase, AlaRS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16, 234734, 292023 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49588, P50475, Q8BGQ7 | |
| Aars | |
| AlaRS Fusion Protein Ag11294 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu