missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Antibody (ALDH1A1/7011R) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP3-14211JF669
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal antibody specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Spécification
| Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
| Recombinant fragment (around aa 315-434) of human Aldehyde Dehydrogenase 1-A1/ALDH1A1 protein (exact sequence is proprietary). (Uniprot: P00352 ) | |
| 0.1 mL | |
| Cancer, Diabetes Research, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers | |
| 216 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| ALDH1A1/7011R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu