missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA515002
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Spécification
ALDH1L1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O75891, P28037, Q8R0Y6 | |
ALDH1L1 | |
KLH conjugated synthetic peptide between 717-746 amino acids from the C-terminal region of human ALDH1L1 | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ALDH1L1 | |
10-formyltetrahydrofolate dehydrogenase; 10fTHF; 10-fTHF; 10FTHFDH; 10-FTHFDH; 1810048F20Rik; aldehyde dehydrogenase 1 family member L1; aldehyde dehydrogenase 1 family, member L1; aldehyde dehydrogenase family 1 member L1; Aldh1l1; Cytosolic 10-formyltetrahydrofolate dehydrogenase; DKFZp781N0997; FBP-CI; FDH; formyltetrahydrofolate dehydrogenase; Fthfd; LOC100855730; MER57B-ALDH1L1; Neut2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
107747, 10840, 64392 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |