missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ALDH4A1 Recombinant Protein
Marque: Abnova™ H00008659-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
AAH07581 | |
Solution | |
8659 | |
ALDH4A1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
ALDH4/P5CD/P5CDh/P5CDhL/P5CDhS | |
ALDH4A1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
87.67 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALDH4A1 | |
Human | |
Yes |