missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aldolase C Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5112299
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Aldolase C Polyclonal Antibody for Western Blot, IP
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008].
Spécification
| Aldolase C | |
| Polyclonal | |
| Unconjugated | |
| ALDOC | |
| AI847350; ALDC; ALDCAA; Aldo3; ALDOC; Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; aldolase, fructose-bisphosphate C; AU040929; Brain-type aldolase; EC 4.1.2.13; F16dip7; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; fructose-bisphosphate aldolase C; RATALDCAA; Scrapie-responsive protein 2; Scrg2; Zebrin II | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 230 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P09972 | |
| ALDOC | |
| E. coli-derived Human Aldolase C/ALDOC fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu