missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALG2 Antibody (OTI3C2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72308APC
This item is not returnable.
View return policy
Description
ALG2 Monoclonal antibody specifically detects ALG2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| ALG2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 85365 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI3C2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| alpha-1,3-mannosyltransferase ALG2, alpha-1,3-mannosyltransferase), asparagine-linked glycosylation 2 homolog (S. cerevisiae, asparagine-linked glycosylation 2 homolog (yeast, asparagine-linked glycosylation 2, alpha-1,3-mannosyltransferase homolog (S.cerevisiae), Asparagine-linked glycosylation protein 2 homolog, CDGII, EC 2.4.1, EC 2.4.1.-, FLJ14511, GDP-Man:Man(1)GlcNAc(2)-PP-dolichol mannosyltransferase, hALPG2, homolog of yeast ALG2, NET38 | |
| Full length human recombinant protein of human ALG2(NP_149078) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction