missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Mouse anti-Human, Clone: ALK/1031, Novus Biologicals™
Mouse Monoclonal Antibody
272.00€
Spécification
| Antigène | ALK/CD246 |
|---|---|
| Clone | ALK/1031 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml |
| Applications | Western Blot |
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Western Blot.Spécification
| ALK/CD246 | |
| 0.2 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 238 | |
| IgG2b κ | |
| Protein A or G purified |
| ALK/1031 | |
| Western Blot 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| Recombinant human ALK/CD246 protein fragment covering ∼aa400-500. | |
| Primary | |
| Store at 4°C. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit